2-Ethylthiophenothiazine ≥97.0% (by total nitrogen basis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
E0447-25G
E0447-500G
TCIAE0447-25GEA
189
DKK
TCIAE0447-25G
TCIAE0447-500G
2-Ethylthiophenothiazine ≥97.0% (by total nitrogen basis)
2-Ethylthiophenothiazine
Formel:
C₁₄H₁₃NS₂ Molekylvægt: 259.39 g/mol |
MDL Number:
MFCD00191362 CAS-nummer: 46815-10-5 |
Specifikationer Testresultater
Appearance | White to Amber to Dark green powder to crystal |
Purity (with Total Nitrogen) | min. 97.0 % |
Melting point | 98.0 to 102.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...