Etilefrine hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Ethylamino-1-(3-hydroxyphenyl)ethanol Hydrochloride

E0381-25G E0381-5G
TCIAE0381-25GEA 2410 DKK
TCIAE0381-25G TCIAE0381-5G
Etilefrine hydrochloride ≥98.0% (by HPLC, titration analysis)
Etilefrine hydrochloride
Formel: C₁₀H₁₅NO₂
Molekylvægt: 217.69 g/mol
MDL Number: MFCD00060167
CAS-nummer: 943-17-9
EINECS: 213-398-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b9a4eb399fb516e1ab63a4791639f25e55e5e33&vid=5b9a4eb399fb516e1ab63a4791639f25e55e5e33&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 117.0 to 123.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR