4-Ethoxy-2-butanone ≥92.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
E0298-25ML
E0298-5ML
TCIAE0298-25MLEA
2710
DKK
TCIAE0298-25ML
TCIAE0298-5ML
4-Ethoxy-2-butanone ≥92.0% (by GC)
4-Ethoxy-2-butanone
Formel:
C₆H₁₂O₂ Kogepunkt: 153°C |
MDL Number:
MFCD00059372 CAS-nummer: 60044-74-8 UN nummer: 1993 ADR: 3,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 92.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...