Ethylenediamine-N,N'-dipropionic acid dihydrochloride ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E0238-1G E0238-25G
TCIAE0238-1GEA 132 DKK
TCIAE0238-1G TCIAE0238-25G
Ethylenediamine-N,N'-dipropionic acid dihydrochloride ≥98.0% (by titrimetric analysis)
Ethylenediamine-N,N'-dipropionic acid dihydrochloride
Formel: C₈H₁₆N₂O₄
MDL Number: MFCD00060168
CAS-nummer: 32705-91-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR