Copper(II) disodium ethylenediaminetetraacetate tetrahydrate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: Ethylenediaminetetraacetic acid Copper(II) disodium salt, Copper(II) disodium 2,2',2'',2'''-(ethane-1,2-diylbis(azanetriyl))tetraacetate tetrahydrate, Copper Disodium Ethylenediaminetetraacetate hydrate

E0090-25G E0090-500G
TCIAE0090-25GEA 132 DKK
TCIAE0090-25G TCIAE0090-500G
Copper(II) disodium ethylenediaminetetraacetate tetrahydrate ≥98.0% (by titrimetric analysis)
Copper(II) disodium ethylenediaminetetraacetate tetrahydrate
Formel: C₁₀H₁₂CuN₂Na₂O₈·4H₂O
Molekylvægt: 469.80 g/mol
MDL Number: MFCD00148920
CAS-nummer: 39208-15-6
EINECS: 254-356-9

Bestil nu

Specifikationer Testresultater

Appearance Blue to Dark blue to Dark purple powder to crystal
Purity (Chelometric Titration) min. 98.0 % (calcd.on anh.substance)
Solubility in Water almost transparency
Water 13.0 to 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR