Ethylenediammonium sulphate ≥98.0% (by titrimetric analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

Synonymer: 1.2-Diaminoethane sulphate

E0082-25G E0082-500G
TCIAE0082-25GEA 132 DKK
TCIAE0082-25G TCIAE0082-500G
Ethylenediammonium sulphate ≥98.0% (by titrimetric analysis)
Ethylenediammonium sulphate
Formel: C₂H₈N₂
MDL Number: MFCD00036417
CAS-nummer: 22029-36-3
EINECS: 244-733-6
UN nummer: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a7abc531d38fdaa452fd50306f3a40d400889cf2&vid=a7abc531d38fdaa452fd50306f3a40d400889cf2&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR