Ethisterone ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

E0040-1G E0040-5G
TCIAE0040-1GEA 140 DKK
TCIAE0040-1G TCIAE0040-5G
Ethisterone ≥98.0% (by HPLC, titration analysis)
Ethisterone
Formel: C₂₁H₂₈O₂
Smeltepunkt: 272°C
MDL Number: MFCD00003656
CAS-nummer: 434-03-7
EINECS: 207-096-5
Merck Index: 14,03741

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D +29.0 to +34.0 deg (C=1, Pyridine)
Solubility in Pyridine almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR