N-Succinimidyl Decanoate ≥97.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,5-Dioxopyrrolidin-1-yl Decanoate, Decanoic acid N-Succinimidyl ester, 1-(Decanoyloxy)-2,5-pyrrolidinedione, Decanoic acid 2,5-Dioxopyrrolidin-1-yl ester

D5833-1G D5833-250MG
TCIAD5833-1GEA 1400 DKK
TCIAD5833-1G TCIAD5833-250MG
N-Succinimidyl Decanoate ≥97.0% (by GC)
N-Succinimidyl Decanoate
Formel: C₁₄H₂₃NO₄
Opbevaringstemperatur: Fryser
MDL Number: MFCD00673470
CAS-nummer: 22102-66-5

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR