2,6-Diisopropylphenylisothiocyanat ≥95.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D5830-1G
D5830-5G
TCIAD5830-1GEA
222
DKK
TCIAD5830-1G
TCIAD5830-5G
2,6-Diisopropylphenylisothiocyanat ≥95.0% (by GC)
2,6-Diisopropylphenylisothiocyanat
Formel:
C₁₃H₁₇NS Kogepunkt: 145°C |
MDL Number:
MFCD00041341 CAS-nummer: 25343-70-8 UN nummer: 2810 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...