N-[2-(Dimethylamino)ethyl]acrylamide ≥98.0% (by GC, titration analysis) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
D5794-25G
D5794-5G
TCIAD5794-25GEA
3800
DKK
TCIAD5794-25G
TCIAD5794-5G
N-[2-(Dimethylamino)ethyl]acrylamide ≥98.0% (by GC, titration analysis) stabilised
N-[2-(Dimethylamino)ethyl]acrylamide
Formel:
C₇H₁₄N₂O Kogepunkt: 125°C Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00053694 CAS-nummer: 925-76-8 EINECS: 213-124-7 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Neutralization titration) | min. 98.0 % |
Specific gravity (20/20) | 0.9600 to 0.9700 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...