2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine ≥98.0% (by HPLC, total nitrogen)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D5787-25G
D5787-5G
TCIAD5787-25GEA
985
DKK
TCIAD5787-25G
TCIAD5787-5G
2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine ≥98.0% (by HPLC, total nitrogen)
2',3'-Di-O-acetyl-5'-deoxy-5-fluoro-N-(pentyloxycarbonyl)cytidine
Formel:
C₁₉H₂₆FN₃O₈ Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD07369303 CAS-nummer: 162204-20-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (with Total Nitrogen) | min. 98.0 % |
Melting point | 109.0 to 113.0 deg-C |
Specific rotation [a]20/D | +82.0 to +86.0 deg (C=1, CHCl3) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...