4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 1,1'-(4,4'-Biphenyldiyl)bis(1H-imidazole)

D5777-1G D5777-5G
TCIAD5777-1GEA 1130 DKK
TCIAD5777-1G TCIAD5777-5G
4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl ≥98.0% (by HPLC, total nitrogen)
4,4'-Di(1H-imidazol-1-yl)-1,1'-biphenyl
Formel: C₁₈H₁₄N₄
Molekylvægt: 286.34 g/mol
MDL Number: MFCD23103195
CAS-nummer: 855766-92-6

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR