1,3-Dimethyl-1,3-diphenyl-1,3-divinyldisiloxane ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D5557-5G
D5557-25G
TCIAD5557-5GEA
698
DKK
TCIAD5557-5G
TCIAD5557-25G
1,3-Dimethyl-1,3-diphenyl-1,3-divinyldisiloxane ≥98.0% (by GC)
1,3-Dimethyl-1,3-diphenyl-1,3-divinyldisiloxane
Formel:
C₁₈H₂₂OSi₂ Molekylvægt: 310.54 g/mol Kogepunkt: 139°C |
MDL Number:
MFCD00053860 CAS-nummer: 2627-97-6 EINECS: 220-101-5 |
Specifikationer Testresultater
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...