3-[Dimethoxy(methyl)silyl]propyl acrylate ≥95.0% (by GC) stabilised
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
3-(Dimethoxy(methyl)silyl)propyl acrylate, Acrylic acid 3-[Dimethoxy(methyl)silyl]propyl ester
Stabilization:
Stabilised with 4-methoxyphenol (MEHQ)
D5556-5G
D5556-25G
TCIAD5556-5GEA
132
DKK
TCIAD5556-5G
TCIAD5556-25G
3-[Dimethoxy(methyl)silyl]propyl acrylate ≥95.0% (by GC) stabilised
3-[Dimethoxy(methyl)silyl]propyl acrylate
Formel:
C₉H₁₈O₄Si Molekylvægt: 218.32 g/mol Opbevaringstemperatur: Køleskab |
MDL Number:
MFCD00156431 CAS-nummer: 13732-00-8 |
Specifikationer Testresultater
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...