Dolasetron Mesylate monohydrate ≥98.0% (by HPLC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
Synonymer:
rel-(5s,6R,8r,9aS)-3-Oxooctahydro-1H-2,6-methanoquinolizin-8-yl 1H-indole-3-carboxylate methanesulphonate hydrate, Dolasetron methanesulphonate, Dolasetron Mesylate hydrate
D5481-200MG
D5481-50MG
TCIAD5481-200MGEA
4580
DKK
TCIAD5481-200MG
TCIAD5481-50MG
Dolasetron Mesylate monohydrate ≥98.0% (by HPLC)
Dolasetron Mesylate monohydrate
Formel:
C₁₉H₂₀N₂O₃·CH₄O₃S·H₂O |
MDL Number:
MFCD01718979 CAS-nummer: 878143-33-0 Merck Index: 14,03411 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 6.10 to 6.70 % |
Water | 3.0 to 5.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...