6-Dehydronandrolone Acetate ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D5471-1G D5471-5G
TCIAD5471-1GEA 378 DKK
TCIAD5471-1G TCIAD5471-5G
6-Dehydronandrolone Acetate ≥98.0% (by HPLC)
6-Dehydronandrolone Acetate
Formel: C₂₀H₂₆O₃
MDL Number: MFCD00271529
CAS-nummer: 2590-41-2

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Melting point 106.0 to 110.0 deg-C
Specific rotation [a]20/D -37.0 to -40.0 deg (C=1,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR