2-(Dimethylamino)benzoesyre ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2-Dimethylaminobenzoic acid

D5463-5G D5463-25G
TCIAD5463-5GEA 1880 DKK
TCIAD5463-5G TCIAD5463-25G
2-(Dimethylamino)benzoesyre ≥98.0% (by HPLC)
2-(Dimethylamino)benzoesyre
Formel: C₉H₁₁NO₂
Molekylvægt: 165.19 g/mol
MDL Number: MFCD00016496
CAS-nummer: 610-16-2
EINECS: 210-209-0

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR