N,N-Dimethyl-1H-imidazole-1-sulphonamide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D5437-1G D5437-5G
TCIAD5437-1GEA 247 DKK
TCIAD5437-1G TCIAD5437-5G
N,N-Dimethyl-1H-imidazole-1-sulphonamide ≥98.0% (by GC)
N,N-Dimethyl-1H-imidazole-1-sulphonamide
Formel: C₅H₉N₃O₂S
Kogepunkt: 100°C
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00955713
CAS-nummer: 78162-58-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=23149f7962b35c3d4f5f6909117de36c7b2e91aa&vid=23149f7962b35c3d4f5f6909117de36c7b2e91aa&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 46.0 to 50.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR