2-(9,9-Dimethyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 9,9-Dimethyl-9H-fluorene-4-boronic acid Pinacol ester, 9,9-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene

D5408-1G D5408-200MG
TCIAD5408-1GEA 2360 DKK
TCIAD5408-1G TCIAD5408-200MG
2-(9,9-Dimethyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥98.0% (by HPLC, titration analysis)
2-(9,9-Dimethyl-9H-fluoren-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel: C₂₁H₂₅BO₂
MDL Number: MFCD30187267
CAS-nummer: 1365692-79-0

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 140.0 to 144.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR