Diethylene glycol diacrylate ≥75.0% (by GC) stabilised

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig


Stabilization: Stabilised with HQ + MEHQ

D5381-25G D5381-100G
TCIAD5381-25GEA 312 DKK
TCIAD5381-25G TCIAD5381-100G
Diethylene glycol diacrylate ≥75.0% (by GC) stabilised
Diethylene glycol diacrylate
Formel: C₁₀H₁₄O₅
Kogepunkt: 96°C
Flammepunkt: 78°C
MDL Number: MFCD00014939
CAS-nummer: 4074-88-8
EINECS: 223-791-6
UN nummer: 2810
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ba227bfe57d152682a7b6677120e90ba026ff80f&vid=ba227bfe57d152682a7b6677120e90ba026ff80f&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 75.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR