9,9-Dimethyl-10-phenyl-9,10-dihydroacridine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D5354-200MG D5354-1G
TCIAD5354-200MGEA 682 DKK
TCIAD5354-200MG TCIAD5354-1G
9,9-Dimethyl-10-phenyl-9,10-dihydroacridine ≥98.0% (by GC)
9,9-Dimethyl-10-phenyl-9,10-dihydroacridine
Formel: C₂₁H₁₉N
MDL Number: MFCD01851187
CAS-nummer: 717880-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=59591745af175c92dec8975a1d04a0ac81cbc0ff&vid=59591745af175c92dec8975a1d04a0ac81cbc0ff&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR