Dienogest ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17alpha-17-Hydroxy-3-oxo-19-norpregna-4,9-diene-21-nitrile, 17-Cyanomethyl-17-hydroxy-estra-4,9-dien-3-one

D5230-250MG D5230-50MG
TCIAD5230-250MGEA 6240 DKK
TCIAD5230-250MG TCIAD5230-50MG
Dienogest ≥98.0% (by HPLC)
Dienogest
Formel: C₂₀H₂₅NO₂
Opbevaringstemperatur: Køleskab
MDL Number: MFCD00868356
CAS-nummer: 65928-58-7
Merck Index: 14,03106

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 4.20 to 4.80 %
Melting point 213.0 to 217.0 deg-C
Specific rotation [a]20/D -302 to -312 deg (C=1, CH2Cl2)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR