3,3-Difluorocyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Farlig

D5225-200MG D5225-1G
TCIAD5225-200MGEA 271 DKK
TCIAD5225-200MG TCIAD5225-1G
3,3-Difluorocyclobutanecarboxylic acid ≥98.0% (by GC, titration analysis)
3,3-Difluorocyclobutanecarboxylic acid
Formel: C₅H₆F₂O₂
Molekylvægt: 136.1 g/mol
MDL Number: MFCD08705857
CAS-nummer: 107496-54-8
UN nummer: 3261
ADR: 8 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6ff006aa50e3ec65d0817a66487543acf687c183&vid=6ff006aa50e3ec65d0817a66487543acf687c183&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 51.0 to 55.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR