1,3-Dibenzoyloxybenzen ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D5207-5G D5207-25G
TCIAD5207-5GEA 165 DKK
TCIAD5207-5G TCIAD5207-25G
1,3-Dibenzoyloxybenzen ≥98.0% (by GC)
1,3-Dibenzoyloxybenzen
Formel: C₂₀H₁₄O₄
MDL Number: MFCD00016576
CAS-nummer: 94-01-9
EINECS: 202-294-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=46fc6f2478b7fcbde014865f3b0126e6c2fd6e96&vid=46fc6f2478b7fcbde014865f3b0126e6c2fd6e96&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR