Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D5176-200MG D5176-1G
TCIAD5176-200MGEA 329 DKK
TCIAD5176-200MG TCIAD5176-1G
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate ≥98.0% (by GC)
Dimethyl (3,3-Difluoro-2-oxoheptyl)phosphonate
Formel: C₉H₁₇F₂O₄P
Kogepunkt: 110°C
MDL Number: MFCD11707108
CAS-nummer: 50889-46-8

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR