1,3-Diallylurea ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D5170-1G D5170-5G
TCIAD5170-1GEA 723 DKK
TCIAD5170-1G TCIAD5170-5G
1,3-Diallylurea ≥98.0% (by GC)
1,3-Diallylurea
Formel: C₇H₁₂N₂O
Kogepunkt: 56°C
MDL Number: MFCD00008639
CAS-nummer: 1801-72-5
EINECS: 217-291-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=405c40e1dd84fbc8b2d5529b42bb8120db2ef5b3&vid=405c40e1dd84fbc8b2d5529b42bb8120db2ef5b3&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 96.0 to 100.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR