2,6-Difluor-3-pyridinboronsyrepinacolester ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,6-Difluoropyridine-3-boronic acid, pinacol ester, 2,6-Difluoropyridine-3-boronic acid Pinacol ester, 2,6-Difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 2-(2,6-Difluoro-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

D5142-200MG D5142-1G
TCIAD5142-200MGEA 165 DKK
TCIAD5142-200MG TCIAD5142-1G
2,6-Difluor-3-pyridinboronsyrepinacolester ≥98.0% (by GC, titration analysis)
2,6-Difluor-3-pyridinboronsyrepinacolester
Formel: C₁₁H₁₄BF₂NO₂
Molekylvægt: 241.04 g/mol
MDL Number: MFCD09037475
CAS-nummer: 1072945-00-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a1945ad1fe659e98d30a13305595c4f34c04c7c6&vid=a1945ad1fe659e98d30a13305595c4f34c04c7c6&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 54.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR