2,6-Difluoro-3-iodopyridine ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D5141-1G
D5141-5G
TCIAD5141-1GEA
526
DKK
TCIAD5141-1G
TCIAD5141-5G
2,6-Difluoro-3-iodopyridine ≥98.0% (by GC)
2,6-Difluoro-3-iodopyridine
Formel:
C₅H₂F₂IN Molekylvægt: 240.98 g/mol |
MDL Number:
MFCD09743595 CAS-nummer: 685517-67-3 |
|
Specifikationer Testresultater
Appearance | White to Light yellow powder to lump |
Purity (GC) | min. 98.0 % |
Melting point | 38.0 to 42.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...