trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (trans,trans)-4-(2,3-Difluoro-4-methylphenyl)-4'-ethyl-1,1'-bi(cyclohexane)

D5127-1G D5127-5G
TCIAD5127-1GEA 608 DKK
TCIAD5127-1G TCIAD5127-5G
trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(2,3-Difluoro-4-methylphenyl)-4'-ethylbicyclohexyl
Formel: C₂₁H₃₀F₂
MDL Number: MFCD26143186
CAS-nummer: 174350-08-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b54d7f25e5bda7b4ecf0e470d042e34212f58567&vid=b54d7f25e5bda7b4ecf0e470d042e34212f58567&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Mesomorphic range 55.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR