9,9-Dimethylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
9,9-Dimethylfluorene-2-boronic acid Pinacol ester, 9,9-Dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl)-9H-fluorene, 2-(9,9-Dimethyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
D5018-1G
D5018-5G
TCIAD5018-1GEA
222
DKK
TCIAD5018-1G
TCIAD5018-5G
9,9-Dimethylfluorene-2-boronic acid pinacol ester ≥98.0% (by HPLC, titration analysis)
9,9-Dimethylfluorene-2-boronic acid pinacol ester
Formel:
C₂₁H₂₅BO₂ |
MDL Number:
MFCD08704229 CAS-nummer: 569343-09-5 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Melting point | 134.0 to 138.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...