2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by HPLC, titration analysis)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
(9,9-Diphenyl-9H-fluoren-2-yl)boronic acid Pinacol ester, 9,9-Diphenyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-fluorene
D5009-200MG
D5009-1G
TCIAD5009-200MGEA
206
DKK
TCIAD5009-200MG
TCIAD5009-1G
2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane ≥97.0% (by HPLC, titration analysis)
2-(9,9-Diphenyl-9H-fluoren-2-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Formel:
C₃₁H₂₉BO₂ |
MDL Number:
MFCD29089356 CAS-nummer: 462128-39-8 |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Neutralization titration) | min. 97.0 % |
Melting point | 198.0 to 202.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...