(R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D4992-1G D4992-5G
TCIAD4992-1GEA 501 DKK
TCIAD4992-1G TCIAD4992-5G
(R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine ≥98.0% (by HPLC, total nitrogen)
(R)-(-)-(3,5-Dioxa-4-phospha-cyclohepta[2,1-a;3,4-a']dinaphthalen-4-yl)dimethylamine
Formel: C₂₂H₁₈NO₂P
MDL Number: MFCD03426988
CAS-nummer: 157488-65-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93f02edb1da1286970d946a353612e20b1620e97&vid=93f02edb1da1286970d946a353612e20b1620e97&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Purity (HPLC) min. 98.0 area%
Optical purity (LC) min. 98.0 ee%
Purity (Nonaqueous Titration) min. 98.0 %
Specific rotation [a]20/D -560 to -600 deg (C=0.3,CHCL3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR