4,4-Diphenylcyclohexanol ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

D4973-1G D4973-5G
TCIAD4973-1GEA 838 DKK
TCIAD4973-1G TCIAD4973-5G
4,4-Diphenylcyclohexanol ≥98.0% (by GC)
4,4-Diphenylcyclohexanol
Formel: C₁₈H₂₀O
Molekylvægt: 252.36 g/mol
MDL Number: MFCD00087400
CAS-nummer: 42420-85-9

Bestil nu

Specifikationer Testresultater

Purity (GC) min. 98.0 %
Melting point 141.0 to 145.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR