Dibromopyromellitic Dianhydride ≥98.0% (by HPLC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4947-1G D4947-5G
TCIAD4947-1GEA 1250 DKK
TCIAD4947-1G TCIAD4947-5G
Dibromopyromellitic Dianhydride ≥98.0% (by HPLC)
Dibromopyromellitic Dianhydride
Formel: C₁₀Br₂O₆
Smeltepunkt: 275°C
MDL Number: MFCD01100205
CAS-nummer: 24848-78-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3cc8ab42b87dacd8eaeae7f83954bafb5e1f2fba&vid=3cc8ab42b87dacd8eaeae7f83954bafb5e1f2fba&prid=web&clid=SAPDAM

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Methanolysis Method) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR