Difluoromethyl phenyl sulphide ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4938-1G D4938-5G
TCIAD4938-1GEA 1180 DKK
TCIAD4938-1G TCIAD4938-5G
Difluoromethyl phenyl sulphide ≥98.0% (by GC)
Difluoromethyl phenyl sulphide
Formel: C₇H₆F₂S
Kogepunkt: 63°C
MDL Number: MFCD16301013
CAS-nummer: 1535-67-7
UN nummer: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e7fe26b4f13cea42d58679ed07d2b5d88c4bdc5&vid=7e7fe26b4f13cea42d58679ed07d2b5d88c4bdc5&prid=web&clid=SAPDAM

Bestil nu


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR