trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
trans-4'-(3,4-Difluorophenyl)-[1,1'-bi(cyclohexan)]-4-one
D4911-1G
D4911-5G
TCIAD4911-1GEA
394
DKK
TCIAD4911-1G
TCIAD4911-5G
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one ≥98.0% (by GC)
trans-4'-(3,4-Difluorophenyl)bicyclohexyl-4-one
Formel:
C₁₈H₂₂F₂O |
MDL Number:
MFCD26143179 CAS-nummer: 147622-85-3 |
|
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 75.0 to 79.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...