4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4897-200MG D4897-1G
TCIAD4897-200MGEA 1310 DKK
TCIAD4897-200MG TCIAD4897-1G
4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene ≥98.0% (by GC)
4,4-Difluoro-1,3-dimethyl-4-bora-3a,4a-diaza-s-indacene
Formel: C₁₁H₁₁BF₂N₂
Smeltepunkt: 138°C
MDL Number: MFCD29472525
CAS-nummer: 154793-49-4

Bestil nu

Specifikationer Testresultater

Appearance Orange to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 144.0 to 148.0 deg-C
Lambda max. 495.0 to 500.0 nm (CH2Cl2)
Molar extinction coefficient min. 55000 (CH2Cl2, 495.0 to 500.0nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR