trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: (trans,trans)-4-(3,4-Difluorophenyl)-4'-vinyl-1,1'-bi(cyclohexane)

D4872-1G D4872-5G
TCIAD4872-1GEA 312 DKK
TCIAD4872-1G TCIAD4872-5G
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl ≥98.0% (by GC)
trans,trans-4-(3,4-Difluorophenyl)-4'-vinylbicyclohexyl
Formel: C₂₀H₂₆F₂
MDL Number: MFCD09842838
CAS-nummer: 142400-92-8

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 44.0 to 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR