5-(Diethylamino)pentyl alcohol ≥98.0% (by GC, titration analysis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4865-1ML D4865-5ML
TCIAD4865-1MLEA 165 DKK
TCIAD4865-1ML TCIAD4865-5ML
5-(Diethylamino)pentyl alcohol ≥98.0% (by GC, titration analysis)
5-(Diethylamino)pentyl alcohol
Formel: C₉H₂₁NO
Kogepunkt: 217°C
Massefylde: 0,88
MDL Number: MFCD00046024
CAS-nummer: 2683-57-0

Bestil nu

Specifikationer Testresultater

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR