11,12-Dihydro-11-phenylindolo[2,3-a]carbazole ≥97.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4860-200MG D4860-1G
TCIAD4860-200MGEA 600 DKK
TCIAD4860-200MG TCIAD4860-1G
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole ≥97.0% (by HPLC, total nitrogen)
11,12-Dihydro-11-phenylindolo[2,3-a]carbazole
Formel: C₂₄H₁₆N₂
MDL Number: MFCD22581305
CAS-nummer: 1024598-06-8

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (with Total Nitrogen) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR