2,3-Dimethoxybenzonitril ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Farlig
|
D4853-25G
D4853-5G
TCIAD4853-25GEA
1170
DKK
TCIAD4853-25G
TCIAD4853-5G
2,3-Dimethoxybenzonitril ≥98.0% (by GC)
2,3-Dimethoxybenzonitril
Formel:
C₉H₉NO₂ |
MDL Number:
MFCD00001784 CAS-nummer: 5653-62-3 EINECS: 227-097-4 UN nummer: 3439 ADR: 6.1,III |
Specifikationer Testresultater
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 44.0 to 48.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...