2',6'-Dimethoxyacetophenon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 1-(2,6-Dimethoxyphenyl)ethan-1-one, 1-(2,6-Dimethoxyphenyl)ethanone

D4849-1G D4849-5G
TCIAD4849-1GEA 132 DKK
TCIAD4849-1G TCIAD4849-5G
2',6'-Dimethoxyacetophenon ≥98.0% (by GC)
2',6'-Dimethoxyacetophenon
Formel: C₁₀H₁₂O₃
Kogepunkt: 136°C
MDL Number: MFCD00008729
CAS-nummer: 2040-04-2
EINECS: 218-034-1

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 68.0 to 72.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR