2,7-Dibromo-9-phenylcarbazole ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

Synonymer: 2,7-Dibromo-9-phenyl-9H-carbazole

D4832-200MG D4832-1G
TCIAD4832-200MGEA 271 DKK
TCIAD4832-200MG TCIAD4832-1G
2,7-Dibromo-9-phenylcarbazole ≥98.0% (by GC)
2,7-Dibromo-9-phenylcarbazole
Formel: C₁₈H₁₁Br₂N
MDL Number: MFCD28147705
CAS-nummer: 444796-09-2

Bestil nu

Specifikationer Testresultater

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 179.0 to 183.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR