2',5'-Dimethoxypropiophenone ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
1-(2,5-Dimethoxyphenyl)-1-propanone
D4817-200MG
D4817-1G
TCIAD4817-200MGEA
312
DKK
TCIAD4817-200MG
TCIAD4817-1G
2',5'-Dimethoxypropiophenone ≥98.0% (by GC)
2',5'-Dimethoxypropiophenone
Formel:
C₁₁H₁₄O₃ Kogepunkt: 168°C |
MDL Number:
MFCD00027013 CAS-nummer: 5803-30-5 EINECS: 227-357-7 |
Specifikationer Testresultater
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...