2,4-Dichloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4805-1G D4805-5G
TCIAD4805-1GEA 394 DKK
TCIAD4805-1G TCIAD4805-5G
2,4-Dichloro-6-phenyl-1,3,5-triazine ≥98.0% (by GC)
2,4-Dichloro-6-phenyl-1,3,5-triazine
Formel: C₉H₅Cl₂N₃
Kogepunkt: 136°C
MDL Number: MFCD00047340
CAS-nummer: 1700-02-3
EINECS: 216-928-6

Bestil nu

Specifikationer Testresultater

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point gt. 119.0 to 123.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR