3',4'-Dimethoxyflavon ≥98.0% (by GC)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 2-(3,4-Dimethoxyphenyl)-4-benzopyrone, 2-(3,4-Dimethoxyphenyl)-4H-chromen-4-one, 2-(3,4-Dimethoxyphenyl)chromen-4-one

D4794-200MG D4794-1G
TCIAD4794-200MGEA 444 DKK
TCIAD4794-200MG TCIAD4794-1G
3',4'-Dimethoxyflavon ≥98.0% (by GC)
3',4'-Dimethoxyflavon
Formel: C₁₇H₁₄O₄
MDL Number: MFCD00143009
CAS-nummer: 4143-62-8

Bestil nu

Specifikationer Testresultater

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 151.0 to 155.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR