N,N'-Dibutylquinacridone ≥98.0% (by HPLC, total nitrogen)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Synonymer: 5,12-Dibutyl-5,12-dihydroquino[2,3-b]acridine-7,14-dione

D4780-200MG D4780-1G
TCIAD4780-200MGEA 649 DKK
TCIAD4780-200MG TCIAD4780-1G
N,N'-Dibutylquinacridone ≥98.0% (by HPLC, total nitrogen)
N,N'-Dibutylquinacridone
Formel: C₂₈H₂₈N₂O₂
Molekylvægt: 424.54 g/mol
Smeltepunkt: 265°C
MDL Number: MFCD12407159
CAS-nummer: 99762-80-8

Bestil nu

Specifikationer Testresultater

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Lambda max. 518.0 to 523.0 nm (CH2Cl2)
Molar extinction coefficient min. 17000 (CH2Cl2, 518.0 to 523.0nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR