Dibenzothiophene-2-carboxaldehyde ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
Synonymer:
Dibenzo[b,d]thiophene-2-carbaldehyde
D4777-1G
D4777-5G
TCIAD4777-1GEA
1320
DKK
TCIAD4777-1G
TCIAD4777-5G
Dibenzothiophene-2-carboxaldehyde ≥98.0% (by GC)
Dibenzothiophene-2-carboxaldehyde
Formel:
C₁₃H₈OS Molekylvægt: 212.27 g/mol |
MDL Number:
MFCD02251670 CAS-nummer: 22099-23-6 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystaline |
Purity (GC) | min. 98.0 % |
Melting point | 105.0 to 109.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...