3,3'-Diethylthiatricarbocyanine iodide ≥98.0% (by total nitrogen basis)

Leverandør: TCI
Ratings: Ingen bedømmelse

Samlet vurdering
Gennemsntilig vurdering

Advarsel

D4773-1G D4773-5G
TCIAD4773-1GEA 493 DKK
TCIAD4773-1G TCIAD4773-5G
3,3'-Diethylthiatricarbocyanine iodide ≥98.0% (by total nitrogen basis)
3,3'-Diethylthiatricarbocyanine iodide
Formel: C₂₅H₂₅IN₂S₂
Smeltepunkt: 211°C
MDL Number: MFCD00041994
CAS-nummer: 3071-70-3
EINECS: 221-342-9

Bestil nu

Specifikationer Testresultater

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Læs mere About VWR