Dibenzo[b,d]furan-4-carbaldehyde ≥98.0% (by GC)
Leverandør: TCI
Ratings:
Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
|
Advarsel
|
D4771-1G
D4771-5G
TCIAD4771-1GEA
337
DKK
TCIAD4771-1G
TCIAD4771-5G
Dibenzo[b,d]furan-4-carbaldehyde ≥98.0% (by GC)
Dibenzo[b,d]furan-4-carbaldehyde
Formel:
C₁₃H₈O₂ |
MDL Number:
MFCD03306027 CAS-nummer: 96706-46-6 |
Specifikationer Testresultater
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 95.0 to 99.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...